Merck
The sensitivity of sheep to vecuronium: an example of the limitations of extrapolation.
Canadian journal of anaesthesia = Journal canadien d'anesthesie (2012-04-06)
Manuel Martin-Flores, Monique D Paré, Luis Campoy, Robin D Gleed
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vecuroniumbromid
Vecuroniumbromid, European Pharmacopoeia (EP) Reference Standard