Merck
  • Crypteins derived from the mouse neuropeptide FF (NPFF)A precursor display NPFF-like effects in nociceptive tests in mice.
Crypteins derived from the mouse neuropeptide FF (NPFF)A precursor display NPFF-like effects in nociceptive tests in mice.
Peptides (2012-05-15)
Jolanta H Kotlinska, Ewa Gibula-Bruzda, Piotr Suder, Magdalena Wasielak, Lauriane Bray, Hana Raoof, Anna Bodzon-Kulakowska, Jerzy Silberring
ZUSAMMENFASSUNG
NPFF precursor, pro-NPFF(A) contains three known bioactive sequences: NPFF (FLFQPQRF-NH(2)), neuropeptide AF (NPAF; AGEGLSSPFWSLAAPQRF-NH(2)) and neuropeptide SF (NPSF; SLAAPQRF-NH(2)). The key-feature of these fragments is their common PQRF-amidated sequence at their C termini. Here, we evaluated the biological activity of two other sequences derived from the mouse NPFF(A) precursor, that does not have PQRF-amidated C-terminus. One peptide was residing between positions 85 and 99 in the mice pro-NPFF(A). This peptide was referred to as neuropeptide SA (NPSA; SAWGSWSKEQLNPQA), assigned due to its flanking amino acids. Another sequence used in the experiments was N-terminal fragment of NPSA, here referred to as neuropeptide SS (NPSS; SAWGSWS). These two peptides, classified as crypteins, were synthesized and tested in the hot-plate and tail immersion tests in mice for their pharmacological activity in morphine-induced antinociception. The effects of both crypteins were compared to NPFF. Our experiments indicated that both crypteins inhibited morphine antinociception and their effects were reversed by RF9, an antagonist of NPFF receptors. These data show that NPSA and NPSS possess NPFF-like anti-opioid activity in these behavioral tests.