Merck
  • Tomatidine promotes the inhibition of 24-alkylated sterol biosynthesis and mitochondrial dysfunction in Leishmania amazonensis promastigotes.
Tomatidine promotes the inhibition of 24-alkylated sterol biosynthesis and mitochondrial dysfunction in Leishmania amazonensis promastigotes.
Parasitology (2012-06-22)
J M Medina, J C F Rodrigues, W De Souza, G C Atella, H Barrabin
ZUSAMMENFASSUNG
Leishmaniasis is a set of clinically distinct infectious diseases caused by Leishmania, a genus of flagellated protozoan parasites, that affects ~12 million people worldwide, with ~2 million new infections annually. Plants are known to produce substances to defend themselves against pathogens and predators. In the genus Lycopersicon, which includes the tomato, L. esculentum, the main antimicrobial compound is the steroidal glycoalkaloid α-tomatine. The loss of the saccharide side-chain of tomatine yields the aglycone tomatidine. In the present study, we investigated the effects of tomatidine on the growth, mitochondrial membrane potential, sterol metabolism, and ultrastructure of Leishmania amazonensis promastigotes. Tomatidine (0·1 to 5 μM) inhibited parasite growth in a dose-dependent manner (IC(50)=124±59 nM). Transmission electron microscopy revealed lesions in the mitochondrial ultrastructure and the presence of large vacuoles and lipid storage bodies in the cytoplasm. These structural changes in the mitochondria were accompanied by an effective loss of mitochondrial membrane potential and a decrease in ATP levels. An analysis of the neutral lipid content revealed a large depletion of endogenous 24-alkylated sterols such as 24-methylene-cholesta-5, 7-dien-3β-ol (5-dehydroepisterol), with a concomitant accumulation of cholesta-8, 24-dien-3β-ol (zymosterol), which implied a perturbation in the cellular lipid content. These results are consistent with an inhibition of 24-sterol methyltransferase, an important enzyme responsible for the methylation of sterols at the 24 position, which is an essential step in the production of ergosterol and other 24-methyl sterols.