Merck
  • Pharmacokinetic analysis of ziconotide (SNX-111), an intrathecal N-type calcium channel blocking analgesic, delivered by bolus and infusion in the dog.
Pharmacokinetic analysis of ziconotide (SNX-111), an intrathecal N-type calcium channel blocking analgesic, delivered by bolus and infusion in the dog.
Neuromodulation : journal of the International Neuromodulation Society (2012-07-04)
Tony L Yaksh, Annelies de Kater, Robin Dean, Brookie M Best, George P Miljanich
ZUSAMMENFASSUNG
Ziconotide is a peptide that blocks N-type calcium channels and is antihyperalgesic after intrathecal (IT) delivery. We here characterize the spinal kinetics of IT bolus and infused ziconotide in dog. Male beagle dogs (N= 5) were prepared with chronic IT lumbar injection and cerebrospinal fluid (LCSF) sampling catheters connected to vest-mounted pumps. Each dog received the following: 1) IT bolus ziconotide (10 µg + 1 µCi (3) H-inulin); 2) IT infusion for 48 hours of ziconotide (1 µg/100 µL/hour); 3) IT infusion for 48 hours of ziconotide (5 µg/100 µL/hour); and 4) intravenous injection of ziconotide (0.1 mg/kg). After IT bolus, LCSF ziconotide and inulin showed an initial peak and biphasic (distribution/elimination) clearance (ziconotide T(1/2-α/β) = 0.14 and 1.77 hours, and inulin T(1/2-α/β) = 0.16 and 3.88 hours, respectively). The LCSF : plasma ziconotide concentration ratio was 20,000:1 at 30 min and 30:1 at eight hours. IT infusion of 1 and then 5 µg/hour resulted in LCSF concentrations that peaked by eight hours and remained stable at 343 and 1380 ng/mL, respectively, to the end of the 48-hour infusions. Terminal elimination T(1/2) after termination of continuous infusion was 2.47 hours. Ziconotide LCSF : cisternal CSF : plasma concentration ratios after infusion of 1 and 5 µg/hour were 1:0.017:0.001 and 1:0.015:0.003, respectively. IT infusion of ziconotide at 1 µg/hour inhibited thermal skin twitch by 24 hours and produced modest trembling, ataxia, and decreased arousal. Effects continued through the 48-hour infusion period, increased in magnitude during the subsequent 5 µg/hour infusion periods, and disappeared after drug clearance. After IT bolus or infusion, ziconotide displays linear kinetics that are consistent with a hydrophilic molecule of approximately 2500 Da that is cleared slightly more rapidly than inulin from the LCSF. Behavioral effects were dose dependent and reversible.