Merck
  • Clinical and forensic examinations of glycemic marker 1,5-anhydroglucitol by means of high performance liquid chromatography tandem mass spectrometry.
Clinical and forensic examinations of glycemic marker 1,5-anhydroglucitol by means of high performance liquid chromatography tandem mass spectrometry.
Forensic science international (2012-07-04)
Cornelius Hess, Bernd Stratmann, Wulf Quester, Burkhard Madea, Frank Musshoff, Diethelm Tschoepe
ZUSAMMENFASSUNG
Postmortem diagnosis of diabetes and a diabetic coma can be difficult because of the lack of characteristic morphological findings. 1,5-Anhydroglucitol (1,5-AG), the 1-deoxy form of glucose, competes with glucose for reabsorption in the kidneys. Therefore, diabetics with a permanent hyperglycemia show significantly lower serum concentrations of 1,5-AG than non-diabetics. A liquid chromatography-mass spectrometric method for the determination of 1,5-AG in serum and postmortem blood was developed and validated according to international guidelines. Linearity was given between 1 μg/ml and 50 μg/ml. Recovery rates ranged between 70.8% and 89.8%, the limit of quantification of the procedure was 0.20 μg/ml, limit of quantification was 0.55 μg/ml. Serum of 199 diabetics and 116 non-diabetics and femoral blood of 31 diabetic and 27 non-diabetic deceased was measured. Average concentrations were significantly (p<0.001) higher in non-diabetics compared to diabetics ante and postmortem. Seven of the diabetics may have died because of a hyperglycemic coma indicated by a sum formula of Traub>450 mg/dl. 1,5-AG average concentrations in these deceased were not significantly different to diabetics which did not die because of a diabetic coma. Concentrations of 1,5-AG give a hint for not well controlled diabetes antemortem and postmortem and can be assumed as an additional and alternative information postmortem to the measurement of HbA1c or fructosamine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,5-Anhydro-D-sorbitol, crystalline, ≥98.0% (TLC)