- Quinone-dependent alcohol dehydrogenases and FAD-dependent alcohol oxidases.
Quinone-dependent alcohol dehydrogenases and FAD-dependent alcohol oxidases.
Biochemistry. Biokhimiia (2012-08-07)
A R Gvozdev, I A Tukhvatullin, R I Gvozdev
PMID22860906
ZUSAMMENFASSUNG
This review considers quinone-dependent alcohol dehydrogenases and FAD-dependent alcohol oxidases, enzymes that are present in numerous methylotrophic eu- and prokaryotes and significantly differ in their primary and quaternary structure. The cofactors of the enzymes are bound to the protein polypeptide chain through ionic and hydrophobic interactions. Microorganisms containing these enzymes are described. Methods for purification of the enzymes, their physicochemical properties, and spatial structures are considered. The supposed mechanism of action and practical application of these enzymes as well as their producers are discussed.