Merck
  • Acacetin protects dopaminergic cells against 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine-induced neuroinflammation in vitro and in vivo.
Acacetin protects dopaminergic cells against 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine-induced neuroinflammation in vitro and in vivo.
Biological & pharmaceutical bulletin (2012-08-07)
Hyo Geun Kim, Mi Sun Ju, Sang Keun Ha, Hyangsook Lee, Hyejung Lee, Sun Yeou Kim, Myung Sook Oh
ZUSAMMENFASSUNG
Acacetin (5,7-dihydroxy-4'-methoxyflavone), a constituent of flavone naturally present in plants, has anti-cancer and anti-inflammatory activities. Neuroinflammation is thought to be one of the major pathological mechanisms responsible for Parkinson's disease (PD), and has been a primary target in the development of treatment for PD. In the present study, we evaluated the neuroprotective effect of acacetin in PD induced by 1-methyl-4-phenylpyridine (MPP+)/or 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) and examined the related pathways in vitro and in vivo. In primary mesencephalic culture, acacetin protected dopaminergic (DA) cells and inhibited production of inflammatory factors such as nitric oxide, prostaglandin E2, and tumor necrosis factor-α against MPP+-induced toxicity in a dose-dependent manner. Then, we confirmed the effect of acacetin (10 mg/kg/d for 3 d, per os (p.o.)) in a mouse model of PD induced by MPTP (30 mg/kg/d for 5 d, intraperitoneally (i.p.)). In the behavioral test (pole test), the acacetin-treated mice showed decreased time of turning and locomotor activity, which were longer in MPTP-only treated mice. In addition, the acacetin-treated group inhibited degeneration of DA neurons and depletion of dopamine level induced by MPTP toxicity in the substantia nigra and striatum of the brain. Moreover, the acacetin-treated group inhibited microglia activation, accompanied by production of inducible nitric oxide synthases and cyclooxygenase-2. These results suggest that acacetin can protect DA neurons against the neurotoxicity involved in PD via its anti-inflammatory action.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acacetin, ≥97.0% (HPLC)