Merck
Serotonin-induced brain glycogenolysis in rainbow trout (Oncorhynchus mykiss).
The Journal of experimental biology (2012-08-10)
Jorge J Pérez-Maceira, María J Mancebo, Manuel Aldegunde
ZUSAMMENFASSUNG
In this study, we evaluated the serotonin-mediated control of cerebral glycogen levels in the rainbow trout, Oncorhynchus mykiss. Intracerebroventricular (i.c.v.) administration of serotonin (5-HT) to normoglycemic trout (time and dose response) decreased glycogen levels in the brain and increased brain glycogen phosphorylase activity (time response). In hypoglycemic fish (that had been fasted for 5 and 10 days), there was a time-dependent decrease in brain glycogen levels; under these conditions, i.c.v. administration of 5-HT also reduced the brain glycogen content in fish that had been fasted for 5 days. In fish with local cerebral hypoglycemia (induced by 2-DG administration), the glycogen levels decreased and, as above, i.c.v. administration of 5-HT also lowered the glycogen content. In hyperglycemic fish, 5-HT did not affect glycogen levels. Administration of receptor agonists 5-HT1A (8-OH-DPAT), 5-HT1B (anpirtoline and CP93129) or 5-HT2 (α-m-5-HT) decreased the brain glycogen levels. This effect was antagonized by the administration of receptor antagonists 5-HT1A (WAY100135 and NAN190), 5-HT1B (NAS181) and 5-HT2B/C (SB206553). Administration of the receptor agonists (±)-DOI (5-HT2A/2C), m-CPP (5-HT2B/2C), BW723C86 (5-HT2B) and WAY 161503 (5-HT2C) led to decreases in the levels of brain glycogen. We found that 5-HT is involved in the modulation of brain glycogen homeostasis in the rainbow trout, causing a glycogenolytic effect when fish are in a normoglycemic or hypoglycemic state, but not when they are in a hyperglycemic state. 5-HT1A, 5-HT1B, 5HT2B and 5-HT2C-like receptors appeared to be involved in the glycogenolytic action of 5-HT, although the effect mediated by 5-HT1A or 5-HT1B was apparently stronger.