Merck
  • Design, synthesis and evaluation of clinafloxacin triazole hybrids as a new type of antibacterial and antifungal agents.
Design, synthesis and evaluation of clinafloxacin triazole hybrids as a new type of antibacterial and antifungal agents.
Bioorganic & medicinal chemistry letters (2012-08-14)
Yan Wang, Guri L V Damu, Jing-Song Lv, Rong-Xia Geng, Da-Cheng Yang, Cheng-He Zhou
ZUSAMMENFASSUNG
A series of clinafloxacin triazole hybrids as a new type of antibacterial and antifungal agents were synthesized for the first time and screened for their antimicrobial efficacy against four Gram-positive bacteria, four Gram-negative bacteria and two fungi by two fold serial dilution technique. The bioactive assay indicated that most of the target compounds displayed broad antimicrobial spectrum and good antibacterial and antifungal activities with low MIC values ranging from 0.25 to 2 μg/mL against all the tested strains which exhibited comparable or even better efficiency in comparison with the reference drugs Chloramphenicol, Clinafloxacin and Fluconazole, respectively. Notably, some synthesized clinafloxacin triazoles showed stronger efficacy against methicillin-resistant Staphylococcus aureus than their parent Clinafloxacin.