Merck
  • Ultrasonic emulsification of food-grade nanoemulsion formulation and evaluation of its bactericidal activity.
Ultrasonic emulsification of food-grade nanoemulsion formulation and evaluation of its bactericidal activity.
Ultrasonics sonochemistry (2012-09-08)
Vijayalakshmi Ghosh, Amitava Mukherjee, Natarajan Chandrasekaran
ZUSAMMENFASSUNG
Basil oil (Ocimum basilicum) nanoemulsion was formulated using non-ionic surfactant Tween80 and water by ultrasonic emulsification method. Process of nanoemulsion development was optimized for parameters such as surfactant concentration and emulsification time to achieve minimum droplet diameter with high physical stability. Surfactant concentration was found to have a negative correlation with droplet diameter, whereas emulsification time had a positive correlation with droplet diameter and also with intrinsic stability of the emulsion. Stable basil oil nanoemulsion with droplet diameter 29.3 nm was formulated by ultrasonic emulsification for 15 min. Formulated nanoemulsion was evaluated for antibacterial activity against Escherichia coli by kinetics of killing experiment. Fluorescence microscopy and FT-IR results showed that nanoemulsion treatment resulted alteration in permeability and surface features of bacterial cell membrane.