- Elastic and bendable caffeine cocrystals: implications for the design of flexible organic materials.
Elastic and bendable caffeine cocrystals: implications for the design of flexible organic materials.
Angewandte Chemie (International ed. in English) (2012-09-13)
Soumyajit Ghosh, C Malla Reddy
PMID22969017
MATERIALIEN