- High-brightness 1.3 μm InAs/GaAs quantum dot tapered laser with high temperature stability.
High-brightness 1.3 μm InAs/GaAs quantum dot tapered laser with high temperature stability.
Optics letters (2012-10-03)
Yulian Cao, Haiming Ji, Pengfei Xu, Yongxian Gu, Wenquan Ma, Tao Yang
PMID23027282
ZUSAMMENFASSUNG
We demonstrate high-brightness 1.3 μm tapered lasers with high temperature stability by using p-doped InAs/GaAs quantum dots (QDs) as the active region. It is found that the beam quality factor M(2) for the devices is almost unchanged as the light power and temperature increase. The almost constant M(2) results from the p-doped QD active region.