Merck
Psychoactive properties of alpha-methyltryptamine: analysis from self reports of users.
Journal of psychoactive drugs (2012-10-16)
James Wilcox
ZUSAMMENFASSUNG
This is a retrospective examination of former users of alpha-methyltryptamine (AMT). A scale of questions was given to subjects who were familiar with the effects of AMT and the outcome was analyzed. Most subjects agreed that AMT had strong hallucinogenic qualities. The most common unwanted effects included anxiety, nausea and moderately severe dysphoria. Several subjects reported significant depression associated with use of AMT. Use of alpha-methyltryptamine could be dangerous in individuals prone to depression.