Merck
Human radiation dose estimation of (11)C-CFT using whole-body PET.
Clinical nuclear medicine (2012-11-17)
Tingting Huang, Hongliang Wang, Ganghua Tang, Xiang Liang, Huaifu Deng, Chang Yi, Xiangsong Zhang
ZUSAMMENFASSUNG
C-Labeled 2-β-carbomethoxy-3-β-(4-fluorophenyl)tropane (C-CFT) is a commonly used positron emission tomography (PET) tracer for dopamine transporters imaging. The present study estimated human radiation absorbed doses of C-CFT based on whole-body PET imaging in healthy subjects. Whole-body PET was performed on 6 subjects after injection of 472.06 ± 116.47 MBq of C-CFT. 7 Frames were acquired for about 70 min in 7 segments of the body. Regions of interest were drawn on PET images of source organs. Residence time was calculated as the area under the time-activity curve. Radiation dosimetry was calculated from organ residence time using the medical internal radiation dosimetry (MIDR) method. The organs with the highest radiation-absorbed doses were the urinary bladder, followed the spleen, pancreas, kidneys, and stomach. The dose-limiting critical organ was the urinary bladder. The effective dose was 8.89E-03 mSv/MBq (22.9 mrem/mCi). Biexponential fitting of mean bladder activity demonstrated that 18% of activity was excreted via the urine. The potential radiation risks of C-CFT associated with in this study are well within accepted limits. C-CFT demonstrates a favorable radiation dose profile in humans and allows multiple PET examinations on the same subject per year.