Merck
  • β-N-Methylamino-L-alanine exposure alters defense against oxidative stress in aquatic plants Lomariopsis lineata, Fontinalis antipyretica, Riccia fluitans and Taxiphyllum barbieri.
β-N-Methylamino-L-alanine exposure alters defense against oxidative stress in aquatic plants Lomariopsis lineata, Fontinalis antipyretica, Riccia fluitans and Taxiphyllum barbieri.
Ecotoxicology and environmental safety (2012-11-28)
Valeska Contardo-Jara, Marc Sebastian Funke, Anja Peuthert, Stephan Pflugmacher
ZUSAMMENFASSUNG
Four different aquatic plants, the Pteridophyte Lomariopsis lineata and the Bryophytes Fontinalis antipyretica, Riccia fluitans and Taxiphyllum barbieri, were tested for their capacity to absorb the neurotoxin β-N-Methylamino-L-alanine (BMAA) from water and thus their possible applicability in a "Green Liver System". After exposure to 10 and 100 μg L(-1) BMAA for 1, 3, 7 and 14 days exposure concentration of medium and tissue were analyzed by LC-MS/MS. The amount removed by the plants within only 1 day was equal to the biological degradation of 14 days. Comparing the "BMAA-removal" capacity of the 4 tested aquatic plants R. fluitans, L. lineata and T. barbieri turned out to be most effective in cleaning the water from this cyanobacterial toxin by up to 97% within 14 days. Activity of the antioxidant enzymes peroxidase (POD) and catalase (CAT), as well as biotransformation enzyme glutathione S-transferase (GST) was compared between exposed and control plants to determine possible harmful effects induced by BMAA. Whereas the Bryophytes displayed increased POD activity and subsequent adaptation when exposed to the lower concentration, as well as partly inhibited antioxidant response at the higher applied BMAA concentration, the Pteridophyte L. lineata reacted with increased POD activity during the whole experiment and increased GST activity after longer exposure for 14 days. To give a recommendation of the suitability of an aquatic plant to be used for sustainable phytoremediation of contaminated water, testing of removal capacity of specific contaminants as well as studying general physiological parameters giving hint on survivability in such environments has to be combined.