Merck
  • Effects of increasing dosages of zeranol implants on lamb growth, carcass characteristics, blood hormones, and nitrogen metabolism.
Effects of increasing dosages of zeranol implants on lamb growth, carcass characteristics, blood hormones, and nitrogen metabolism.
Journal of animal science (2012-12-12)
S R Eckerman, G P Lardy, M M Thompson, M L Van Emon, B W Neville, P T Berg, C S Schauer
ZUSAMMENFASSUNG
The objective of this research was to compare the growth performance, incidence of prolapse and mortality, carcass characteristics, blood hormone concentration, and N balance of lambs implanted with increasing dosages of zeranol. One hundred forty-four crossbred lambs (29.6 ± 4.9 kg) were used in a completely random design and placed into 16 feedlot pens (4 pens/treatment) for a 116 d finishing study. Lambs were fed an 84.7% corn and 15.3% market lamb pellet (DM basis) diet ad libitum. Treatments were 0, 12, 24, and 36 mg zeranol (Ralgro; Schering-Plough), and lambs were implanted in the ear according to treatment on d 0. Lambs were weighed. Thirty lambs (67.6 ± 3.4 kg) and 96 lambs (65.8 ± 5.1 kg) were harvested on d 84 and d 118, respectively. Carcass data were collected 24 h after chill. Blood samples were collected on d 0, 28, 56, 70, 82, 99, and 116 from 64 lambs (29.6 ± 2.1 kg) in the feedlot study (subsample of 4 lambs per pen) and analyzed for thyroxine, triiodothyronine, and IGF-I. A second study was conducted to compare effects of 0, 12, 24, or 36 mg zeranol on N balance in 16 crossbred lambs (34.8 ± 2.1 kg). There were no differences among treatments for BW, ADG, DMI, and G:F (P > 0.05) in the feedlot study. However, there was a linear increase for incidence of prolapse (P = 0.006; 2.78, 5.55, 24.98, and 27.75%, respectively) and mortality (P = 0.005; 0.00, 5.55, 11.10, and 13.88%, respectively) as zeranol dosage increased. Carcass characteristics, blood hormone concentrations, and N balance were not affected by treatment (P > 0.05). These results indicate zeranol increases incidence of prolapse and mortality without increasing growth performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Zearalanol, ~98% (HPLC)