Merck
  • Effects of perinatal daidzein exposure on subsequent behavior and central estrogen receptor α expression in the adult male mouse.
Effects of perinatal daidzein exposure on subsequent behavior and central estrogen receptor α expression in the adult male mouse.
Progress in neuro-psychopharmacology & biological psychiatry (2012-12-27)
Chengjun Yu, Fadao Tai, Shuangyan Zeng, Xia Zhang
ZUSAMMENFASSUNG
Daidzein is one of the most important isoflavones present in soy and it is unique as it can be further metabolized to equol, a compound with greater estrogenic activity than other isoflavones. The potential role of daidzein in the prevention of some chronic diseases has drawn public attention and increased its consumption in human, including in pregnant women and adolescent. It is unclear whether perinatal exposure to daidzein through maternal diets affects subsequent behavior and central estrogen receptor α (ERα) expression in male adults. Following developmental exposure to daidzein through maternal diets during perinatal period, subsequent anxiety-like behavior, social behavior, spatial learning and memory of male mice at adulthood were assessed using a series of tests. The levels of central ER α expression were also examined using immunocytochemistry. Compared with the controls, adult male mice exposed to daidzein during the perinatal period showed significantly less exploration, higher levels of anxiety and aggression. They also displayed more social investigation for females and a tendency to improve spatial learning and memory. The mice with this early daidzein treatment demonstrated significantly higher levels of ERα expression in several brain regions such as the bed nucleus of the stria terminalis, medial preoptic, arcuate hypothalamic nucleus and central amygdaloid mucleus, but decreased it in the lateral septum. Our results indicated that perinatal exposure to daidzein enhanced masculinization on male behaviors which is assocciated with alterations in ERα expression levels led by perinatal daidzein exposure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Supelco
Daidzein, analytical standard