Merck
  • Helium ion microscopy: a new tool for imaging novel mesoporous silica and organosilica materials.
Helium ion microscopy: a new tool for imaging novel mesoporous silica and organosilica materials.
Chemical communications (Cambridge, England) (2013-01-24)
Andrea S Terpstra, Kevin E Shopsowitz, Camille F Gregory, Alan P Manning, Carl A Michal, Wadood Y Hamad, Jijin Yang, Mark J MacLachlan
ZUSAMMENFASSUNG
Helium ion microscopy (HIM) has been used to image mesoporous silica and organosilica for the first time. Images of chiral nematic silica, ethylenesilica, and new benzenesilica reveal the structural organization, pore dimensions and connectivity of these materials on the nanometer length scale.