Merck
  • Cell penetrating peptide tethered bi-ligand liposomes for delivery to brain in vivo: Biodistribution and transfection.
Cell penetrating peptide tethered bi-ligand liposomes for delivery to brain in vivo: Biodistribution and transfection.
Journal of controlled release : official journal of the Controlled Release Society (2013-01-29)
Gitanjali Sharma, Amit Modgil, Buddhadev Layek, Kanwardeep Arora, Chengwen Sun, Benedict Law, Jagdish Singh
ZUSAMMENFASSUNG
Targeted nano-particulate systems hold extraordinary potential for delivery of therapeutics across blood brain barrier (BBB). In this work, we investigated the potential of novel bi-ligand (transferrin-poly-l-arginine) liposomal vector for delivery of desired gene to brain, in vivo. The in vivo evaluation of the delivery vectors is essential for clinical translation. We followed an innovative approach of combining transferrin receptor targeting with enhanced cell penetration to design liposomal vectors for improving the transport of molecules into brain. The biodistribution profile of 1, 1'-dioctadecyl-3,3,3',3'-tetramethyl-indocarbocyanine iodide(DiR)-labeled liposomes was evaluated in adult rats after single intravenous injection at dose of 15.2μmoles of phospholipids/kg body weight. We demonstrated that bi-ligand liposomes accumulated in rat brain at significantly (p<0.05) higher concentrations as compared to the single-ligand (transferrin) or plain liposomes. In addition, the bi-ligand liposomes resulted in increased expression of β-galactosidase(β-gal) plasmid in rat brain tissue in comparison to the single-ligand liposomes. Histological examination of the transfected tissues did not show any signs of tissue necrosis or inflammation. Hemolysis assay further authenticated the biocompatibility of bi-ligand liposomes in blood up to 600 nmoles of phospholipids/1.4×10(7) erythrocytes. The findings of this study provide important and detailed information regarding the distribution of bi-ligand liposomes in vivo and accentuate their ability to demonstrate improved brain penetration and transfection potential over single-ligand liposomes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly-L-Arginin -hydrochlorid, mol wt 5,000-15,000
Sigma-Aldrich
Poly-L-Arginin -hydrochlorid, mol wt 15,000-70,000
Sigma-Aldrich
Poly-L-Arginin -hydrochlorid, mol wt >70,000