Merck
  • Molecular weight distribution analysis by ultracentrifugation: adaptation of a new approach for mucins.
Molecular weight distribution analysis by ultracentrifugation: adaptation of a new approach for mucins.
Carbohydrate polymers (2013-03-08)
Richard B Gillis, Gary G Adams, Bettina Wolf, Monica Berry, Tabot M D Besong, Anthony Corfield, Samil M Kök, Ray Sidebottom, David Lafond, Arthur J Rowe, Stephen E Harding
ZUSAMMENFASSUNG
Mucins are the key macromolecular component of mucus, nature's natural lubricant, and one of the most important physical properties is their molecular weight distribution. A new approach for polydisperse polymers was recently published based on sedimentation velocity in the analytical ultracentrifuge and converts a distribution of sedimentation coefficient g(s) vs. s plot into a distribution of molecular weight utilising the power-law or scaling relationship between the sedimentation coefficient and molecular weight, s=κsMw(b) where s is the sedimentation coefficient, Mw is the weight average molecular weight and κs and b are characteristic coefficients related to conformation. We investigate the possibility of using a large database of previously published values of s an M to define κs and b for both aqueous solution and aqueous solution supplemented by 6M guanidine hydrochloride (a solvent which helps to minimise sample degradation). These values are then applied to a study of the molecular weight distributions of preparations of human gastric mucin in the different solvents and at different stages of purification.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Guanidin -hydrochlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Guanidin -hydrochlorid, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidin -hydrochlorid, ≥98%
Sigma-Aldrich
Guanidin -hydrochlorid -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidin -hydrochlorid -Lösung, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Guanidin -hydrochlorid, ≥99.0% (AT)
Sigma-Aldrich
Guanidin -hydrochlorid, BioUltra, Molecular Biology, ≥99.5% (AT)