Merck
  • Cytochrome P450-derived eicosanoids and vascular dysfunction in coronary artery disease patients.
Cytochrome P450-derived eicosanoids and vascular dysfunction in coronary artery disease patients.
Atherosclerosis (2013-03-08)
Robert N Schuck, Katherine N Theken, Matthew L Edin, Melissa Caughey, Almasa Bass, Kyle Ellis, Bryant Tran, Savanna Steele, Brian P Simmons, Fred B Lih, Kenneth B Tomer, Michael C Wu, Alan L Hinderliter, George A Stouffer, Darryl C Zeldin, Craig R Lee
ZUSAMMENFASSUNG
Accumulating preclinical and epidemiologic evidence has emerged to suggest that modulation of cytochrome P450 (CYP)-mediated eicosanoid metabolism may be a viable vascular protective therapeutic strategy for the secondary prevention of coronary artery disease (CAD). The functional relationship between CYP-derived eicosanoid metabolite levels and vascular dysfunction in humans with established CAD, however, has not been evaluated. Therefore, we characterized the relationship between inter-individual variation in soluble epoxide hydrolase (sEH) and CYP ω-hydroxylase metabolism and established vascular function phenotypes predictive of prognosis in a cohort of patients with atherosclerotic cardiovascular disease. Plasma epoxyeicosatrienoic acid (EET), dihydroxyeicosatrienoic acid (DHET), and 20-hydroxyeicosatetraenoic acid (20-HETE) levels were quantified by HPLC-MS/MS in 106 patients with stable, angiographically-confirmed CAD. Relationships between biomarkers of CYP-mediated eicosanoid metabolism and vascular function phenotypes were evaluated by Pearson's correlation. A significant inverse association was observed between 20-HETE levels (a biomarker of CYP ω-hydroxylase metabolism) and brachial artery flow-mediated dilation (r = -0.255, p = 0.010). An inverse association was also observed between 14,15-EET:DHET ratios (a biomarker of sEH metabolism) and both monocyte chemoattractant protein-1 levels (r = -0.252, p = 0.009) and a consolidated cellular adhesion molecule 'score' reflecting the levels of E-selectin and P-selectin (r = -0.216, p = 0.027). No associations with C-reactive protein or epithelial neutrophil-activating protein-78 levels were observed. Collectively, these findings demonstrate that enhanced CYP ω-hydroxylase and sEH metabolic function are associated with more advanced endothelial dysfunction and vascular inflammation, respectively, in patients with established atherosclerotic cardiovascular disease. These findings lay the foundation for future clinical research in this area.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
20-Hydroxy-(5Z,8Z,11Z,14Z)-eicosatetraenoic acid, ~100 μg/mL in ethanol, ≥90% (HPLC)