Merck
The pentachlorophenol-dehalogenating Desulfitobacterium hafniense strain PCP-1.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences (2013-03-13)
Richard Villemur
ZUSAMMENFASSUNG
In this report, a complete description of Desulfitobacterium hafniense strain PCP-1 is presented. The D. hafniense strain PCP-1 was isolated from a methanogenic consortium for its capacity to dehalogenate pentachlorophenol (PCP) into 3-chlorophenol. This strain is also capable of dehalogenating several other chloroaromatic compounds and tetrachloroethene into trichloroethene. Four gene loci encoding putative chlorophenol-reductive dehalogenases (CprA2 to CprA5) were detected, and the products of two of these loci have been demonstrated to dechlorinate different chlorinated phenols. Strain PCP-1 was used in laboratory-scale bioprocesses to degrade PCP present in contaminated environments. Desulfitobacterium hafniense PCP-1 is an excellent candidate for the development of efficient bioprocesses to degrade organohalide compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pentachlorphenol, 97%
Sigma-Aldrich
3-Chlorphenol, 98%
Supelco
Pentachlorphenol -Lösung, certified reference material, 5000 μg/mL in methanol
Supelco
Pentachlorphenol, analytical standard