Merck
  • Catalytic enantioselective desymmetrization of meso cyclic anhydrides via iridium-catalyzed hydrogenation.
Catalytic enantioselective desymmetrization of meso cyclic anhydrides via iridium-catalyzed hydrogenation.
Organic letters (2013-03-15)
Tang-Lin Liu, Wei Li, Huiling Geng, Chun-Jiang Wang, Xumu Zhang
ZUSAMMENFASSUNG
A novel method to desymmetrize meso-anhydrides into lactones via asymmetric hydrogenation catalyzed by the Ir-C3*-TunePhos complex has been developed. Various chiral lactones were synthesized with full conversion and excellent enantioselectivity under high reaction temperature.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis