Merck
Synthesis and functionalization of [60]fullerene-fused imidazolines.
Organic letters (2013-03-19)
Cheng-Lin He, Rui Liu, Dan-Dan Li, San-E Zhu, Guan-Wu Wang
ZUSAMMENFASSUNG
The silver carbonate promoted reaction of [60]fullerene with (Z)-N-arylbenzamidines afforded the unprecedented C60-fused imidazoline derivatives in high yields. Substrates with both electron-donating and -withdrawing groups on aromatic rings could be employed. In addition, the electrochemistry and further selective functionalization of the obtained C60-fused imidazolines were investigated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Silbercarbonat, 99%
Sigma-Aldrich
Silbercarbonat auf Celit®, extent of labeling: ~50 wt. % loading
Sigma-Aldrich
Silbercarbonat, purum p.a., ≥99.0% (AT)