Merck
Selective adsorption of hemoglobin using polymer-grafted-magnetite nanocellulose composite.
Carbohydrate polymers (2013-03-19)
Thayyath Sreenivasan Anirudhan, Sylaja Raveendran Rejeena
ZUSAMMENFASSUNG
A novel adsorbent, poly(methacrylic acid-co-vinyl sulfonic acid)-grafted-magnetite nanocellulose composite (P(MAA-co-VSA)-g-MNCC) was synthesized for adsorbing hemoglobin (Hb) selectively from aqueous solutions. FTIR, XRD and DLS analyses were carried out to characterize the material. Hb exhibited a decrease in α-helix and an increase in β-sheet structure, upon immobilization onto P(MAA-co-VSA)-g-MNCC. The maximum adsorption was found to be at pH 6.5 with a monolayer capacity of 248.19mg/g at 30°C. Adsorption capacity attained saturation within 2h. The kinetic data were found to follow pseudo-second-order model which is based on chemisorption. Adsorption behavior was observed to be endothermic in nature. P(MAA-co-VSA)-g-MNCC can be used in the selective adsorption of Hb from mixture of proteins. Spent adsorbent was effectively regenerated with 0.01M KOH. Present investigation had shown that P(MAA-co-VSA)-g-MNCC would be a promising material for the selective recovery of Hb from aqueous solutions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Kaliumhydroxid, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Kaliumhydroxid, technical, ≥85%, powder
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Supelco
Kaliumhydroxid-Konzentrat, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
Kaliumhydroxid, tested according to Ph. Eur.
Sigma-Aldrich
Kaliumhydroxid, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 20,000, contains MEHQ as inhibitor