Merck
  • Hetero diels-alder chemistry for the functionalization of single-walled carbon nanotubes with cyclopentadienyl end-capped polymer strands.
Hetero diels-alder chemistry for the functionalization of single-walled carbon nanotubes with cyclopentadienyl end-capped polymer strands.
Macromolecular rapid communications (2013-03-19)
Nicolas Zydziak, Corinna M Preuss, Volker Winkler, Michael Bruns, Christof Hübner, Christopher Barner-Kowollik
ZUSAMMENFASSUNG
Single-walled carbon nanotubes (SWCNTs) are pre-functionalized with a pyridinyl-based dithioester to undergo a hetero Diels-Alder (HDA) reaction with cyclopentadienyl end-capped poly(methyl)methacrylate (Mn = 2700 g mol(-1) , PDI = 1.14). Fourier transform infrared spectroscopy, thermogravimetric analysis, elemental analysis (EA), and X-ray photoelectron spectroscopy (XPS) evidence the success of the grafting process. The estimated resulting grafting density (from XPS and EA) via the HDA reaction increases by a factor of more than two (0.0774 chains·nm(-2) via XPS) compared with typical values obtained via a direct cyclopentadiene driven Diels-Alder conjugation onto non-functional SWCNTs under similar conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Poly(methylmethacrylat), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methylmethacrylat)
Sigma-Aldrich
Poly(methylmethacrylat), average Mw ~15,000 by GPC, powder
Sigma-Aldrich
Poly(methylmethacrylat)
Sigma-Aldrich
Pyridin -hydrochlorid, 98%
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Supelco
Pyridin, analytical standard
Sigma-Aldrich
Poly(methylmethacrylat), isotaktisch, >80% isotactic
Sigma-Aldrich
Pyridin -hydrobromid, 98%
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Pyridin -hydrochlorid, purum, ≥98.0% (AT)
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methylmethacrylat), analytical standard, 2,480,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Pyridin -Lösung, certified reference material, 2000 μg/mL in methanol
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Supelco
Poly(methylmethacrylat), analytical standard, suitable for gel permeation chromatography (GPC), 8,000