- Mutasynthesis of fluorinated pactamycin analogues and their antimalarial activity.
Mutasynthesis of fluorinated pactamycin analogues and their antimalarial activity.
Organic letters (2013-03-26)
Khaled H Almabruk, Wanli Lu, Yuexin Li, Mostafa Abugreen, Jane X Kelly, Taifo Mahmud
PMID23521145
ZUSAMMENFASSUNG
A mutasynthetic strategy has been used to generate fluorinated TM-025 and TM-026, two biosynthetically engineered pactamycin analogues produced by Streptomyces pactum ATCC 27456. The fluorinated compounds maintain excellent activity and selectivity toward chloroquine-sensitive and multidrug-resistant strains of malarial parasites as the parent compounds. The results also provide insights into the biosynthesis of 3-aminobenzoic acid in S. pactum.
MATERIALIEN