Merck
  • Mathematical modeling and finite element simulation of slow release of drugs using hydrogels as carriers with various drug concentration distributions.
Mathematical modeling and finite element simulation of slow release of drugs using hydrogels as carriers with various drug concentration distributions.
Journal of pharmaceutical sciences (2013-03-26)
Yihan Xu, Yuxi Jia, Zhao Wang, Zhaojing Wang
ZUSAMMENFASSUNG
In drug release systems using hydrogels as carriers, the presence of the polymer network will reduce the drug release rate, which can extend the release period. For a controlled-release process of drug, usually the ideal situation is to get a zero-order drug release rate. In this paper, the mathematical model of hydrogel swelling processes is constructed on the basis of a biphasic theory, and then an integrated equation that considers both water convection and drug diffusion phenomena is used to describe the drug release process. The effects of the initial drug concentration with nonuniform distributions along the radial direction of hydrogel carriers on the release of drugs are studied through simulating two-dimensional hydrogel swelling processes by means of the COMSOL Multiphysics software. The simulation results show that along with the hydrogel swelling, the drug release rate is changing, and the major influencing factors of the drug release rate are water convection and drug diffusion coefficient, which are affected by water volume fraction, drug concentration distribution in matrix, and carrier radius. The results also indicate that the initial drug concentration distribution following a sine curve can result in an ideal zero-order release process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylzellulose, viscosity: 4,000 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methylzellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~86,000
Sigma-Aldrich
Methylzellulose, viscosity: 1,500 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methylzellulose, viscosity: 15 cP
Sigma-Aldrich
Methylzellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Methylzellulose, viscosity: 25 cP
Sigma-Aldrich
Methylzellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methylzellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~120,000
Sigma-Aldrich
Methocel® A15 LV, 27.5-31.5% methoxyl basis
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~90,000
Sigma-Aldrich
Methocel® MC, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methylzellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methylzellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP