Merck
  • Synthesis of skeletally diverse and stereochemically complex library templates derived from isosteviol and steviol.
Synthesis of skeletally diverse and stereochemically complex library templates derived from isosteviol and steviol.
Organic letters (2013-03-28)
Oliver E Hutt, Trinh L Doan, Gunda I Georg
ZUSAMMENFASSUNG
We have applied a diversity-oriented approach for the synthesis of skeletally diverse and stereochemically complex templates for small-molecule library production by performing Beckmann rearrangement and Beckmann fragmentation reactions on the bicyclo[3.2.1]octane rings of steviol and isosteviol, aglycones derived from the diterpene natural product stevioside. The optimization of these two reaction pathways is presented along with the successful application of a photo-Beckmann rearrangement. This work also led to the discovery of cyano-Prins-type and Thorpe-Ziegler-type cyclization reactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Sigma-Aldrich
Oktan, reagent grade, 98%
Sigma-Aldrich
Oktan, anhydrous, ≥99%
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Oktan, analytical standard
Sigma-Aldrich
Stevioside, ≥98% (HPLC)
Sigma-Aldrich
Oktan, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
Steviol, ≥98% (HPLC), powder
Supelco
Discovery® Cyano Supelguard Kartusche, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4 mm