Merck
  • Targeting heat shock proteins by phenethyl isothiocyanate results in cell-cycle arrest and apoptosis of human breast cancer cells.
Targeting heat shock proteins by phenethyl isothiocyanate results in cell-cycle arrest and apoptosis of human breast cancer cells.
Nutrition and cancer (2013-03-28)
Ruma Sarkars, Sutapa Mukherjee, Madhumita Roy
ZUSAMMENFASSUNG
Heat shock proteins (HSPs) are chaperones for several client proteins involved in transcriptional regulation, signal transduction, and cell cycle control. HSPs (27, 70, and 90) are abundantly expressed in a wide range of cancers and are transcriptionally regulated by heat shock factor (HSF1). Most of the synthetic HSP inhibitors exhibit toxicity, therefore, searching for inhibitors with limited or no toxicity will be of help. The objective of the present study was to determine the effect of natural isothiocyanate (phenethyl isothiocyanate; PEITC) on different HSPs (27, 70, and 90) and HSF1 in 2 breast cancer cell lines, namely breast adenocarcinoma MCF-7 (with wild type p53) and highly metastatic breast cancer cell MDA-MB-231 (with mutated p53). PEITC significantly inhibited the expression of HSPs (particularly HSP 90) and HSF1. Molecular consequences due to HSP inhibition were downregulation of cell-cycle regulatory proteins like Cyclin B1, CDK1, Cdc25C, PLK-1, and upregulation of p21 irrespective of p53 status. These modulations were accompanied by cell-cycle arrest at G2/M phase and apoptosis by activation of caspases 3 and 9. PEITC therefore may be regarded as a potent HSP inhibitor and an antitumor agent in the treatment of breast cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cytochrom c aus Pferdeherz, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrom c aus Rinderherz, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrom c aus Pferdeherz, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrom c aus Rinderherz, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
Cytochrom c aus Pferdeherz, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrom c aus Pferdeherz, BioReagent, suitable for GFC marker
Sigma-Aldrich
ProteoMass Cytochrom c MALDI-MS-Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Sigma-Aldrich
Cytochrom c aus Taubenbrustmuskel, ≥95% based on Mol. Wt. 12,173 basis