Merck
  • Enantioselective synthesis of (E)-δ-silyl-anti-homoallylic alcohols via an enantiodivergent hydroboration-crotylboration reaction of a racemic allenylsilane.
Enantioselective synthesis of (E)-δ-silyl-anti-homoallylic alcohols via an enantiodivergent hydroboration-crotylboration reaction of a racemic allenylsilane.
Organic letters (2013-03-29)
Ming Chen, William R Roush
ZUSAMMENFASSUNG
The enantioselective hydroboration of racemic allenylsilane (±)-4 with ((d)Ipc)2BH proceeds via enantiodivergent pathways to give vinylborane 11 and crotylborane intermediate (S)-E-5. Subsequent crotylboration of aldehyde substrates with (S)-E-5 at -78 °C provides (E)-δ-silyl-anti-homoallylic alcohols in 71-89% yield and with 93-96% ee.