Merck
Remote manipulation of posterior lamellar corneal grafts using a magnetic field.
Cornea (2013-03-30)
Yoav Nahum, Tilda Barliya, Irit Bahar, Tami Livnat, Yael Nisgav, Dov Weinberger
ZUSAMMENFASSUNG
In posterior lamellar keratoplasty procedures such as Descemet stripping endothelial keratoplasty and Descemet membrane endothelial keratoplasty, the lamellar graft is manipulated directly or by injecting an air bubble. This preliminary study sought to evaluate the feasibility of guiding lamellar corneal grafts by generating a magnetic field. Rabbit and porcine Descemet stripping endothelial keratoplasty and Descemet membrane endothelial keratoplasty grafts were manually produced and immersed in a ferromagnetic solution containing nanomagnetic particles conjugated to streptavidin or in gadoteric acid. For the feasibility study, grafts were transferred to an artificial anterior chamber or plastic test tube and a magnetic field was generated with a handheld NdFeB disc magnet. The presence and the sustainability of graft motion were documented under various conditions. For the semiquantitative study, whole or partial grafts were transferred to a plastic test tube after immersion, and the amount of tissue retraction induced by the remote magnet was graded. The grafts were successfully manipulated in all directions by the magnet, from a distance of up to 7 mm. They remained ferromagnetic more than 24 hours after immersion in the ferromagnetic solutions. The degree of retraction was affected by graft size, immersion time, time from immersion, and immersion solution. Posterior lamellar corneal grafts may be made ferromagnetic and remotely manipulated by creation of a magnetic field. The ferromagnetic properties are adjustable. This technique holds promise in attaching and repositioning grafts during keratoplasty. Further research is needed to assess the possible effects of ferromagnetic solutions on corneal endothelial cells and on lamellar graft clarity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Methyl-D-Glucamin, 99.0-100.5% (titration)
Sigma-Aldrich
N-Methyl-D-Glucamin, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
Meglumin, 99.0-100.5% dry basis, meets USP testing specifications