Merck
Chitosan and oligochitosan enhance the resistance of peach fruit to brown rot.
Carbohydrate polymers (2013-04-03)
Zengxin Ma, Lingyu Yang, Haixia Yan, John F Kennedy, Xianghong Meng
ZUSAMMENFASSUNG
The effects of chitosan and oligachitosan on resistance induction of peach fruit against brown rot caused by Monilinia fructicola were investigated. Both chitosan and oligochitosan showed significant effect on controlling this disease. Moreover, chitosan and oligochitosan delayed fruit softening and senescence. The two antifungal substances enhanced antioxidant and defense-related enzymes, such as catalase (CAT), peroxidase (POD), β-1,3-glucanase (GLU) and chitinase (CHI), and they also stimulated the transcript expression of POD and GLU. These findings suggest that the effects of chitosan and oligochitosan on disease control and quality maintenance of peach fruit may be associated with their antioxidant property and the elicitation of defense responses in fruit.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chitin aus Garnelenschalen, practical grade, powder
Sigma-Aldrich
Chitin aus Garnelenschalen, BioReagent, suitable for analysis of chitinase, purified powder