Merck
  • Preparation of entangled nanocellulose fibers from APMP and its magnetic functional property as matrix.
Preparation of entangled nanocellulose fibers from APMP and its magnetic functional property as matrix.
Carbohydrate polymers (2013-04-03)
Wei Li, Xin Zhao, Shouxin Liu
ZUSAMMENFASSUNG
Nanocellulose fibers (NCFs) aerogels were prepared from poplar alkaline peroxide mechanical pulp (APMP) using physical ultrasonication method. As raw materials, the unique mechanical effects of APMP cause the fiber folding and loose during the pulping process, which was beneficial to further chemical purification and subsequent treatment for long and entangled NCFs. The obtained NCFs exhibited higher crystallinity (77.8%) compared with that of APMP (72.6%) together with diameters range from 20 to 90 nm and self-assembled to network. The primary thermal degradation of NCFs occurred at 331.5°C. The prepared NCFs network aerogels acted as matrix which can prevent the growth and aggregation of ferromagnetic CoFe2O4 nanoparticles. The ratio of the CoFe2O4 of magnetic composites increased from 34 wt% to 75 wt%, and the magnetic properties were all increased with increasing the reaction concentration of FeSO4/CoCl2 salt.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, ReagentPlus®, ≥99%
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Cobalt(II)-chlorid, 97%
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Cobalt(II)-chlorid, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
Cobaltchlorid 0,1 mol/l -Lösung
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Cobalt(II)-chlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Cobalt(II)-chlorid, AnhydroBeads, −10 mesh, 99.995% trace metals basis