Merck
Effects of extractives on some properties of bagasse/high density polypropylene composite.
Carbohydrate polymers (2013-04-03)
Shabnam Sheshmani
ZUSAMMENFASSUNG
In this study, the effects of two variable parameters, namely the extractives and filler loading level, on the physical properties of composites were examined. Composites based on high density polyethylene (HDPE), bagasse flour (BF) as filler were made by injection molding. In order to increase the interphase adhesion, maleic anhydride grafted polyethylene (MAPE) was added as a coupling agent to all the composites studied. Three different solvents, ethanol-benzene, 1% NaOH and hot-water, were used to remove extractives. Physical properties, namely, water absorption (WA) and thickness swelling (TS) were investigated for a long period. At same filler loading, composites made with extracted bagasse had higher WA and TS values. In addition, the TS of samples showed a similar pattern to the water uptake data. The difference in WA between extracted and unextracted composites is due to blocking of -OH groups by extractives. The results also showed that as the BF content was increased, significant increase in WA and TS occurred. Statistical analysis confirmed that the effects of both variables and their interactions on the WA and TS properties were significant at 1% confidence level.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Supelco
Benzol, analytical standard
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Polyethylen, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Supelco
Benzol -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methanol
Sigma-Aldrich
Polyethylen, low density, melt index 25 g/10 min (190°C/2.16kg)
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylen, High density, melt index 12 g/10 min (190 °C/2.16kg)
Polyethylen (LDPE), ERM®, certified reference material
Sigma-Aldrich
Polyethylen, Medium density
Sigma-Aldrich
Polyethylen, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylen, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Supelco
Polyethylen, analytical standard, suitable for gel permeation chromatography (GPC), 2,000