Merck
Biological activities of selected basidiomycetes from Yemen.
Die Pharmazie (2013-04-06)
M Al-Fatimi, G Schröder, H Kreisel, U Lindequist
ZUSAMMENFASSUNG
In a previous paper we demonstrated the results of biological screening of Yemeni basidiomycetes. The present study was aimed to investigate the antimicrobial and the antioxidant activity of further basidiomycetes collected in Yemen. Dichloromethane, methanol and aqueous extracts of the fruiting bodies of 25 species were screened in vitro for their antibacterial activities against three Gram-positive bacteria (Staphyloccocus aureus, Bacillus subtilis, Micrococcus flavus) and two Gram-negative bacteria (Escherichia coli, Pseudomonas aeruginosa), against six human fungal pathogens (Candida albicans, Candida krusei, Aspergillus fumigatus, Mucor sp., Microsporum gypseum, Trichophyton mentagrophytes) and against one non human pathogenic fungus (Candida maltosa). The results indicated that 75 extracts exhibited activity against one or more of the bacteria. The methanol extracts of Agaricus cf. bernardii, Agrocybe pediades, Chlorophyllum molybdites, Coriolopsis polyzona, Ganoderma xylonoides, Pycnoporus sanguineus, Trametes lactinea and Trametes cingulata showed activity against all tested bacteria. The highest antibacterial activity was exhibited by methanol extracts from Chlorophyllum molybdites, Ganoderma xylonoides and Trametes cingulata and Agaricus cf. bernardii, Agrocybe pediades, Coriolopsis polyzona, Pycnoporus sanguineus and Trametes lactinea. The methanol extracts of Chlorophyllum molybdites, Ganoderma xylonoides and Pycnoporus sanguineus showed considerable antifungal activities against the tested fungal strains. Strong antioxidative effects employing the DPPH assay were exhibited by methanol extracts from Chlorophyllum molybdites, Ganoderma xylonoides, Hexagonia velutina, Pycnoporus sanguineus, Trametes lactinea and Trametes cingulata. Our previous and presented studies about 48 basidiomycetes collected in Yemen provide evidence that basidiomycetes from the Arabic region so far should attract more attention as potential source for new biologically active agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl