Merck
  • Exhaled breath condensate formate after inhaled allergen provocation in atopic asthmatics in vivo.
Exhaled breath condensate formate after inhaled allergen provocation in atopic asthmatics in vivo.
The Journal of asthma : official journal of the Association for the Care of Asthma (2013-04-06)
Roby Greenwald, Brent A Johnson, Aimee Hoskins, Ryszard Dworski
ZUSAMMENFASSUNG
The dual actions of S-nitrosoglutathione reductase comprise reduction of S-nitrosoglutathione, a potent endogenous airway smooth muscle relaxant that is depleted in asthmatics, and detoxification of formaldehyde to formate. Airway formate production is increased in children with asthma, suggesting increased activity of S-nitrosoglutathione reductase. We determined formate in exhaled breath condensate from adult atopic asthmatics with asthma exacerbation produced by inhaled allergen in vivo, Twenty-two adult atopic asthmatics underwent inhaled allergen challenge using specific allergen. Exhaled breath condensate was collected at baseline, 1 h after inhalation of the provocative dose of allergen, and then every 2 h for 8 h during the challenge. Formate was analyzed by ion chromatography, Eleven asthmatics developed an isolated early airway response, and another 11 volunteers early response followed by late airway response (dual response). Formate concentrations doubled 1 h post-challenge in asthmatics with dual-airway response but essentially unchanged in patients with an isolated early reaction, Dual-airway response to allergen in atopic asthmatics could be associated with increased activity of S-nitrosoglutathione reductase as suggested by greater concentrations of formate in exhaled breath condensate. Measurement of formate in exhaled breath condensate could serve as a noninvasive biomarker of S-nitrosoglutathione reductase activity in vivo. Our results need to be confirmed in a larger group of asthmatics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetyl-β-Methylcholin-Chlorid, ≥98% (TLC), powder