Merck
  • Production of mouse anti-quail IgY and subsequent labeling with horseradish peroxidase using cyanuric chloride.
Production of mouse anti-quail IgY and subsequent labeling with horseradish peroxidase using cyanuric chloride.
Journal of microbiology and biotechnology (2013-04-10)
Neema Kassim, Adelard B Mtenga, Won-Bo Shim, Duck-Hwa Chung
ZUSAMMENFASSUNG
Polyclonal antibodies labeled with a tracer have been commonly used as secondary antibodies in immunochemical assays to quantify the concentration of antibody-antigen complexes. The majority of these antibodies conjugated with a tracer are commercially available, with the exception of few untouched targets. This study focused on the production and application of mouse anti-quail IgY as an intermediate antibody to link between quail egg yolk IgY and goat anti-mouse IgG-HRP as primary and secondary antibodies, respectively. Subsequently, the produced mouse anti-quail IgY was labeled with horseradish peroxidase (HRP) and its efficiency on enzyme linked immunosorbent assay (ELISA) was compared with that of commercial rabbit anti-chicken IgY-HRP. As an intermediate antibody, mouse anti-quail IgY was successfully produced with good affinity and sensitivity (1:10,000) to the primary and secondary antibodies. Subsequently, mouse anti-quail IgY was effectively conjugated with HRP enzyme, resulting in a secondary antibody with good sensitivity (1:10,000) to quail anti-V. parahaemolyticus and V. vulnificus IgY. The detection limit was 10(5) CFU/ml for both V. parahaemolyticus and V. vulnificus. The efficiency of the produced conjugate to detect quail IgY on ELISA was comparable to that of the commercial rabbit anti-chicken IgY-HRP, and hence the produced and labeled mouse anti-quail IgY-HRP can be used as a secondary antibody to detect any antibody produced in quail.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyanurchlorid, 99%