Merck
  • Damage investigation on tungsten and diamond diffractive optics at a hard x-ray free-electron laser.
Damage investigation on tungsten and diamond diffractive optics at a hard x-ray free-electron laser.
Optics express (2013-04-11)
Fredrik Uhlén, Daniel Nilsson, Anders Holmberg, Hans M Hertz, Christian G Schroer, Frank Seiboth, Jens Patommel, Vivienne Meier, Robert Hoppe, Andreas Schropp, Hae Ja Lee, Bob Nagler, Eric Galtier, Jacek Krzywinski, Harald Sinn, Ulrich Vogt
ZUSAMMENFASSUNG
Focusing hard x-ray free-electron laser radiation with extremely high fluence sets stringent demands on the x-ray optics. Any material placed in an intense x-ray beam is at risk of being damaged. Therefore, it is crucial to find the damage thresholds for focusing optics. In this paper we report experimental results of exposing tungsten and diamond diffractive optics to a prefocused 8.2 keV free-electron laser beam in order to find damage threshold fluence levels. Tungsten nanostructures were damaged at fluence levels above 500 mJ/cm(2). The damage was of mechanical character, caused by thermal stress variations. Diamond nanostructures were affected at a fluence of 59 000 mJ/cm(2). For fluence levels above this, a significant graphitization process was initiated. Scanning Electron Microscopy (SEM) and µ-Raman analysis were used to analyze exposed nanostructures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wolfram, powder, ≤10 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Diamant, nanopowder, <10 nm particle size (TEM), ≥97% trace metals basis
Sigma-Aldrich
Wolfram, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Wolfram, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Diamant, nanopowder, <10 nm particle size (TEM), ≥95% trace metals basis
Sigma-Aldrich
Diamant, synthetic monocrystalline powder, ≤1 μm
Sigma-Aldrich
Wolfram, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Wolfram, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Wolfram, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Wolfram, foil, thickness 0.127 mm, ≥99.9% trace metals basis