Merck
Controlled cobalt doping in biogenic magnetite nanoparticles.
Journal of the Royal Society, Interface (2013-04-19)
J M Byrne, V S Coker, S Moise, P L Wincott, D J Vaughan, F Tuna, E Arenholz, G van der Laan, R A D Pattrick, J R Lloyd, N D Telling
ZUSAMMENFASSUNG
Cobalt-doped magnetite (CoxFe3 -xO4) nanoparticles have been produced through the microbial reduction of cobalt-iron oxyhydroxide by the bacterium Geobacter sulfurreducens. The materials produced, as measured by superconducting quantum interference device magnetometry, X-ray magnetic circular dichroism, Mössbauer spectroscopy, etc., show dramatic increases in coercivity with increasing cobalt content without a major decrease in overall saturation magnetization. Structural and magnetization analyses reveal a reduction in particle size to less than 4 nm at the highest Co content, combined with an increase in the effective anisotropy of the magnetic nanoparticles. The potential use of these biogenic nanoparticles in aqueous suspensions for magnetic hyperthermia applications is demonstrated. Further analysis of the distribution of cations within the ferrite spinel indicates that the cobalt is predominantly incorporated in octahedral coordination, achieved by the substitution of Fe(2+) site with Co(2+), with up to 17 per cent Co substituted into tetrahedral sites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Kobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Kobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Kobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Kobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Kobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Kobalt, rod, diam. 5.0 mm, 99.95% trace metals basis