- Resolution of enantiomers of 19-hydroxyeicosatetraenoate and 18-hydroxyeicosatetraenoate by chiral phase high-performance liquid chromatography of naphthoyl ester derivatives.
Resolution of enantiomers of 19-hydroxyeicosatetraenoate and 18-hydroxyeicosatetraenoate by chiral phase high-performance liquid chromatography of naphthoyl ester derivatives.
Journal of chromatography (1990-04-06)
E H Oliw
PMID2361991