- [Septic arthritis caused by Streptococcus suis].
[Septic arthritis caused by Streptococcus suis].
Ugeskrift for laeger (2013-06-01)
Sofie Sommer Hedegaard, Matthias Zaccarin, Jens Lindberg
PMID23721843
ZUSAMMENFASSUNG
Streptococcus suis is a global endemic swine pathogen. S. suis can cause meningitis, endocarditis and severe sepsis in humans, who are exposed to swine. Human infection with S. suis was first reported in 1968, since then, human infections have been sporadic although an outbreak in China counted 215 cases. In a rare case of disseminated arthritis we found that correct clinical diagnosis was difficult due to unspecific symptomatology and slow growing bacterial culture. However, conducting thorough examinations is crucial, and if treated correctly the outcome is favourable.