Merck
  • Comparison of intratympanic methylprednisolone and gentamicin for Meniere's disease may be misleading.
Comparison of intratympanic methylprednisolone and gentamicin for Meniere's disease may be misleading.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery (2013-07-03)
Benjamin T Crane
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6α-Methylprednisolon, ≥98%
Methylprednisolon, European Pharmacopoeia (EP) Reference Standard