Merck
[Pitfalls in the interpretation of laboratory parameters-electrolytes, urea, creatinine].
Therapeutische Umschau. Revue therapeutique (2013-07-24)
Lorenz Risch, Bernhard Hess
ZUSAMMENFASSUNG
When it comes to interpret parameters of electrolyte balance and kidney function, it is important to keep pathophysiology and the theory on reference intervals in mind. Hyponatremia is most often caused by excess water. A low sodium concentration in urine should prompt a clinical evaluation of volume status. In case of suspected acute kidney failure, fractionated sodium excretion and fractionated urea excretion are able to provide insights on prerenal or renal origin of the disorder. Disruption in potassium homoeostasis can occur due to changes in supply or renal elimination as well as due to changes in the potassium balance between the extra- and intracellular compartments. The transtubular potassium gradient can help in the differential diagnosis of hyperkalemia. Evaluation of kidney function should begin with determination of serum creatinine, accompanied by an estimate of the glomerular filtration rate, as calculated by the CKD-EPI equation. As a consequence of non-renal determinants of serum creatinine, this equation has been shown to overestimate true GFR in elderly and hospitalized patients. This can result in overdosing of renally-cleared drugs. Clearance determinations can be of use in this context.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Harnstoff, analytical standard
Sigma-Aldrich
Harnstoff, meets USP testing specifications
USP
Harnstoff, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Millipore
Harnstoff -Lösung, suitable for microbiology, 40% in H2O
Harnstoff, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C