Merck
  • LPS and HIV gp120 modulate monocyte/macrophage CYP27B1 and CYP24A1 expression leading to vitamin D consumption and hypovitaminosis D in HIV-infected individuals.
LPS and HIV gp120 modulate monocyte/macrophage CYP27B1 and CYP24A1 expression leading to vitamin D consumption and hypovitaminosis D in HIV-infected individuals.
European review for medical and pharmacological sciences (2013-07-24)
M R Pinzone, M Di Rosa, B M Celesia, F Condorelli, M Malaguarnera, G Madeddu, F Martellotta, D Castronuovo, M Gussio, C Coco, F Palermo, S Cosentino, B Cacopardo, G Nunnari
ZUSAMMENFASSUNG
Vitamin D deficiency is very common among HIV-infected subjects. We cross-sectionally evaluated the prevalence and risk factors for hypovitaminosis D in 91 HIV-infected Italian patients. We studied in a cohort of 91 HIV-infected Italian patients the metabolism of Vitamin D by evaluating the in vitro expression of CYP27B1, CYP24A1 and vitamin D receptor (VDR) by monocytes and macrophages stimulated with the viral envelope protein gp120 or lipopolysaccharide (LPS). The prevalence of vitamin D deficiency (25OHD < 10 ng/ml) and vitamin D insufficiency (25OHD 10-30 ng/ml) was 31% and 57%, respectively. In univariate analysis, female sex (p = 0.01), increasing age (p = 0.05), higher highly sensitive-C reactive protein (p = 0.025), higher parathyroid hormone (PTH) (p = 0.043) and lower BMI (p = 0.04) were associated with vitamin D deficiency. In multivariate analysis, the association was still significant only for PTH (p = 0.03) and female sex (p = 0.03). Monocyte stimulation with LPS (100 ng/ml) or gp120 (1 µg/ml) significantly upregulated CYP27B1 mRNA expression. Moreover, gp120 significantly increased VDR mRNA levels. On the contrary, neither LPS nor gp120 modified CYP24A1 levels. Macrophage stimulation with LPS (100 ng/ml) significantly upregulated CYP27B1 and CYP24A1 mRNA expression. When monocytes were cultured in the presence of 25OHD (40 ng/ml) and stimulated with LPS we detected significantly lower levels of 25OHD in the supernatant. Vitamin D deficiency was very common in our cohort of HIV-infected patients. Chronic inflammation, including residual viral replication, may contribute to hypovitaminosis D, by modulating vitamin D metabolism and catabolism. Systematic screening may help identifying subjects requiring supplementation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
25-Hydroxyvitamin D2, ≥95.0% (HPLC)
Supelco
25-Hydroxyvitamin D2 -Lösung, 50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®