Merck
Metallic nanoparticles and their medicinal potential. Part I: gold and silver colloids.
Therapeutic delivery (2013-07-26)
Leena Loomba, Tiziano Scarabelli
ZUSAMMENFASSUNG
The article highlights the synthesis, properties and recent advances in therapeutic possibilities of metallic nanoparticles. Nanometallic structures that behave as drug-carrying agents, gene regulators, imaging agents and photoresponsive assemblies have been discussed in the context of cells and many debilitating diseases. These structures not only act as alternatives to molecule-based systems, but also possess new physical and chemical properties, which confer substantive advantages in medicinal field.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Silber, conductive paste
Sigma-Aldrich
Silber, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silber, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silber, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silber, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silber, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Silber-Nanodrähte, diam. × L 200  nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silber, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Goldbeschichteter Glasobjektträger, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Silber, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silber, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silber, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silber, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silber, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silber, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Silber, colloidal, 65-75% Ag basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Silber, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Goldbeschichteter Glasobjektträger, layer thickness 1000 Å, 99.999% (Au)
Silber, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95+%
Gold, rod, 50mm, diameter 3.0mm, as drawn, 99.95%
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis