Merck
  • Light microscopical localization of enzymes by means of cerium-based methods. I. Detection of acid phosphatase by a new cerium-lead-technique (Ce-Pb-method).
Light microscopical localization of enzymes by means of cerium-based methods. I. Detection of acid phosphatase by a new cerium-lead-technique (Ce-Pb-method).
Acta histochemica (1985-01-01)
N Zimmermann, K J Halbhuber
ZUSAMMENFASSUNG
Cerium-III-ions are more and more used as capturing reagent and opaque marker for the electron microscopic localization of a number of H2O2-generating enzymes as well as phosphohydrolases. Contrary to its advantages over common lead methods in the histochemical detection of enzyme activities at the electron microscopic level, cerium-based methods proved to be a failure for light microscopic investigations. Therefore, our cerium-based method for the ultrahistochemical detection of acid phosphatase was developed for further observations at the light microscopic level. The principle of that new Ce-Pb-method is the conversion of light microscopic not visible cerium phosphate into lead phosphate by the secondary capture reagent alkaline lead citrate. Finally, the lead phosphate can be visualized as lead sulfide in the section. The Ce-Pb-method in its finally proposed manner was compared with a common lead method and showed a range of advantages. Because of that fact, the new Ce-Pb-method is recommended for a broader use in histochemistry, e.g. for the light microscopic enzyme investigation parallel to ultrahistochemical preparations.