Merck
  • Modulation of adjuvant-enhanced delayed-type hypersensitivity by the interferon inducers poly I:C and Newcastle disease virus.
Modulation of adjuvant-enhanced delayed-type hypersensitivity by the interferon inducers poly I:C and Newcastle disease virus.
International archives of allergy and applied immunology (1986-01-01)
C A Kraaijeveld, W Kamphuis, B J Benaissa-Trouw, M Harmsen, H Snippe
ZUSAMMENFASSUNG
The modulation by the interferon (IFN) inducers poly I:C and Newcastle disease virus (NDV) of the effector phase of adjuvant-enhanced delayed-type hypersensitivity (DH) was studied in mice. A strongly enhanced DH was induced in mice to ultraviolet (UV) light inactivated Semliki forest virus (SFV) by the use of the adjuvant dimethyldioctadecylammonium bromide. At day 6 after intracutaneous immunization, DH was elicited with SFV and measured 24 and 48 h later as increase in footpad thickness (footpad swelling test). Systemic, intravenous administration of either poly I:C, UV-inactivated NDV, or NDV-induced IFN prior to elicitation of DH with antigen resulted in a temporarily suppressed DH reaction. Both the poor swelling at 3 h and strong swelling at 24 h were suppressed, while the swelling at 48 h was enhanced. The model described provides a sensitive in vivo method to study modulating effects of drugs and microbial agents on the effector phase of DH.