Merck
  • In vivo inhibition of chicken brain acetylcholinesterase and neurotoxic esterase in relation to the delayed neurotoxicity of leptophos and cyanofenphos.
In vivo inhibition of chicken brain acetylcholinesterase and neurotoxic esterase in relation to the delayed neurotoxicity of leptophos and cyanofenphos.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer (1986-09-01)
S A Soliman, A Curley, J Farmer, R Novak
ZUSAMMENFASSUNG
An equimolal single dose (1 mmole/kg) of leptophos or cyanofenphos was given orally to chickens to assay the clinical and biochemical neurotoxic effects of these two organophosphorus insecticides. Parathion and TOCP at 2 and 1000 mg/kg of chicken body weight were tested in the same manner as negative and positive neurotoxicants, respectively. Three birds of each of five groups tested were sacrificed 1,2,3,7,14,21 and 28 days after treatment and the brains were taken for the biochemical tests. Acetylcholinesterase (AChE) and neurotoxic esterase (NTE) activities were determined in the brain microsomal fractions. In addition, the AChE activity in the brain soluble fractions was measured. Clinical observations indicated that leptophos-, cyanofenphos- and parathion-treated chickens became acutely poisoned but recovered from the typical cholinergic signs in a day or two. However, about 10 to 15 days later leptophos- and cyanofenphos-treated chickens developed the characteristic leg weakness and unrecoverable ataxia seen in birds given TOCP. The biochemical results indicated that cyanofenphos followed by leptophos and parathion produced more in vivo AChE inhibition than that produced by TOCP in both chicken brain soluble and microsomal fractions. Results suggested that there are no correlations between the in vivo effect of TOCP, leptophos and cyanofenphos on AChE and phenyl valerate-total hydrolyzing activities and the ability of these chemicals to produce neuropathy in hens. The results obtained from this study of the in vivo effect of the tested compounds on chicken brain NTE activity present an acceptable correlation between the inhibition of this enzyme and the ability of these chemicals to induce neuropathy. The mechanism and explanation for this correlation are presented. The in vivo effect of the tested compounds on the chicken brain NTE activity was determined using the indirect and a new direct method. The data presented in this report suggested that the new direct technique of assaying NTE activity using 4-nitrophenyl valerate (4-NPV) as substrate, can be useful in the in vivo screening studies of organophosphates for their ability to induce neuropathy in hens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Leptophos, PESTANAL®, analytical standard