Merck
  • 2(3)-tert-butyl-4-hydroxyanisole inhibits oxidative metabolism of aflatoxin B1 in isolated rat hepatocytes.
2(3)-tert-butyl-4-hydroxyanisole inhibits oxidative metabolism of aflatoxin B1 in isolated rat hepatocytes.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.) (1989-10-01)
J J Ch'ih, D W Biedrzycka, T Lin, M O Khoo, T M Devlin
ZUSAMMENFASSUNG
Previous studies indicate that dietary administration of phenolic antioxidants, 2(3)-tert-butyl-4-hydroxyanisole (BHA) and 3,5-di-tert-butyl-4-hydroxytoluene, inhibits the carcinogenic effect of a number of chemical carcinogens including aflatoxin B1 (AFB1). Induction of hepatic enzymes, such as glutathione S-transferase, UDP-glucuronyltransferase, and epoxide hydrolase, has been shown to be responsible for the reduction of AFB1 cytotoxic and carcinogenic effects. The effect of BHA on AFB1 activation was examined in vitro utilizing isolated rat hepatocytes and liver microsomes. In hepatocytes, the total AFB1 content and bound form of AFB1 were 3.4 and 1.4 pmol/10(6) cells, respectively. In the cell-free microsomal activating system, 2.2 pmol were activated per mg of microsomal protein during 60 min of incubation. BHA (0.1-0.5 mM) inhibited AFB1 activation and binding in both systems in a dose-dependent manner; in hepatocytes, 90% inhibition was observed at 0.5 mM. Analyzing various AFB1 adducts, BHA (0.25 mM)-treated hepatocytes contained a significantly reduced amount of AFB1 macromolecular adducts. The antioxidant neither stimulated nor inhibited the cytosolic glutathione S-transferase and microsomal UDP-glucuronyltransferase activities. Analysis of various hydroxylated (aflatoxins M1 and Q1 (AFM1 and AFQ1] and demethylated (aflatoxin P1 (AFP1] metabolites of AFB1 in both the conjugated and unconjugated form indicated that there was a 30-50% reduction of unconjugated AFP1, AFQ1, and AFM1, whereas AFB1 was increased 3-fold. There was no significant change of conjugated metabolites. The effect of BHA on AFB1 activation in hepatocytes was compared with that of other cytochrome P-450 inhibitors; the ED50 values of SKF 525A, BHA, and metyrapone were 9 microM, 40 microM, and 280 microM, respectively. In the cell-free microsomal system, biotransformation of AFB1 to AFP1, AFM1, and AFQ1 was also inhibited. Kinetic analysis of p-nitroanisole O-demethylase activity of rat liver microsomes demonstrated that BHA inhibited noncompetitively with an apparent Ki of 90 microM. In the absence of enzyme induction, the phenolic antioxidant, BHA, blocks the oxidative biotransformation of AFB1 in isolated hepatocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
2-tert-Butyl-4-hydroxyanisol, United States Pharmacopeia (USP) Reference Standard